TeSR™ E7™ Medium for Reprogramming (2-Component)
Fournisseur: STEMCELL Technologies
05914
STMC05914EA 324 EUR InStock
STMC05914
TeSR™ E7™ Medium for Reprogramming (2-Component)
Milieux de culture cellulaire
Feeder-free and animal component-free reprogramming medium for human iPS cell induction.
TeSR™-E7™ (2-component) is an animal component-free and defined reprogramming culture medium optimised for the generation of human iPS cells without the use of feeders. It is based on the E7 formulation.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité