MyoCult™ SF Expansion 10X Supplement (Human)
Fournisseur: STEMCELL Technologies
05982
STMC05982EA 242 EUR InStock
STMC05982
MyoCult™ SF Expansion 10X Supplement (Human)
Milieux de culture cellulaire
Serum-free supplement for the derivation and expansion of human skeletal muscle progenitor cells (myoblasts).
Support the serum-free culture of primary human skeletal muscle progenitor cells and human pluripotent stem cell (hPSC)-derived myoblasts using MyoCult™-SF expansion 10X supplement (Human). Available either individually or as part of the MyoCult™-SF expansion supplement kit (Human), the supplement has a serum-free formulation and must be combined with a basal medium (DMEM with 1000 mg/l D-glucose; sold separately) to prepare MyoCult™-SF expansion medium. This medium is optimised for the derivation and expansion of human skeletal muscle progenitor cells in vitro. If using primary myoblasts, we recommend coating cultureware with MyoCult™-SF attachment substrate. For hPSC-derived myoblasts, use Corning® Matrigel® hESC-qualified matrix. Myoblasts cultured using MyoCult™-SF expansion medium are compatible with MyoCult™ differentiation kit (Human) and can be used for further differention into multinucleated myotubes.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité