MyoCult™ Expansion 10X Supplement (Mouse)
Fournisseur: STEMCELL Technologies
05985
STMC05985EA 125 EUR InStock
STMC05985
MyoCult™ Expansion 10X Supplement (Mouse)
Milieux de culture cellulaire
Serum-containing supplement for the expansion of mouse skeletal muscle progenitor cells (satellite cells).
MyoCult™ Expansion 10X Supplement (Mouse), when combined with DMEM/F-12 to prepare MyoCult™ Expansion Medium, is for the culture of mouse skeletal muscle progenitor cells (satellite cells). MyoCult™ Expansion Medium has been optimised for the expansion of mouse skeletal muscle satellite cells isolated by fluorescence-activated cell sorting (FACS), as well as for culturing single isolated myofibers grown in suspension. Satellite cells cultured using MyoCult™ Expansion Medium can be differentiated into multinucleated myotubes. For preparation of MyoCult™ Expansion Medium, DMEM/F-12 with 15 mm HEPES is required in addition to MyoCult™ Expansion 10X Supplement. For expansion of satellite cells, cultureware must be coated with a matrix such as Corning Matrigel® Basement Membrane Matrix.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité