Carmine alum solution pour colorant
Fournisseur: STEMCELL Technologies
07070
STMC07070EA 369 EUR InStock
STMC07070
Carmine alum solution pour colorant
Carmine alum
Dye for staining mouse mammary gland whole mounts.
Carmine Alum is used for the whole mount staining of mouse mammary gland.
Contains
Carmine and aluminum potassium sulfate in water
ProClin™ is added as a preservative
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité