ITS Supplement-B
Fournisseur: STEMCELL Technologies
07155
STMC07155EA 249 EUR InStock
STMC07155
ITS Supplement-B
Milieux de culture cellulaire
For neural differentiation of mouse ES and iPS cells.
ITS Supplement-B is a medium supplement containing insulin, apo-transferrin (iron-poor), and selenium in phosphate-buffered saline. It is recommended for use in the in vitro differentiation of mouse embryonic stem (ES) cells and mouse induced pluripotent stem (iPS) cells to neural cell types.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité