DisCoZyme 2, ACF
Fournisseur: STEMCELL Technologies
07450
STMC07450EA 468 EUR InStock
STMC07450
DisCoZyme 2, ACF
Suppléments et additifs pour culture cellulaire
Collagenase/dispase blend with high collagenase and high caseinase activity.
  • Obtain cleaner cultures with an animal component-free solution
  • Dissociate tissues efficiently with a collagenase/dispase blend, high caseinase activity
Use DisCoZyme 2, Animal Component-Free (ACF) for tissue dissociation in applications where prevention of the introduction of potential animal-derived pathogens is necessary. This collagenase and dispase blend is sourced from clostridium histolyticum and bacillus polymyxa, respectively, and contains a minimum of 250 collagenase units/mg and 2000 caseinase units/mg of dry weight. Digest native collagen fibrils commonly found in connective tissues while the neutral amino-endoprotease, dispase, cleaves the N-terminal peptide bond of non-polar amino acid residues. The mild proteolytic activity of dispase allows it to maintain membrane integrity while isolating and passaging of primary cells. DisCoZyme 2, ACF is designed to contain high levels of collagenase and caseinase activity and has been used for the digestion of lung (Fujino et al.; Driscoll et al.) and neural tissues (Panchision et al.). Use DisCoZyme 1, ACF, if lower levels of caseinase activity are desired.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité