Trypsin-EDTA (0,25%)
Fournisseur: STEMCELL Technologies
07901
STMC07901EA 97 EUR InStock
STMC07901
Trypsin-EDTA (0,25%)
Suppléments et additifs pour culture cellulaire
Enzymatic cell dissociation reagent.
  • Dissociate cells efficiently with a combined solution
  • EDTA enhances Trypsin-mediated cell detachment
Use Trypsin-EDTA (0,25%) for enzymatic dissociation of adherent cells, cell aggregates and tissues into single-cell suspensions. EDTA weakens cell-cell adhesion, which improves trypsin's access to hydrolysis-targeted peptide bonds, and enhances detachment of adherent cells. Trypsin-EDTA is also available in a lower concentration (0,05%), for applications that require more gentle dissociation.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité