0.1% Gelatin in Water
Fournisseur: STEMCELL Technologies
07903
STMC07903EA 55 EUR InStock
STMC07903
0.1% Gelatin in Water
Suppléments et additifs pour culture cellulaire
Coating for cultureware.
  • A natural biomaterial derived from collagen
  • Gelatin mimics the extracellular matrix
  • Versatile substrate is useful for many routine cell culture applications
Support the growth of mouse embryonic stem cells (mESCs), mouse induced pluripotent stem cells (miPSCs), or primary embryonic fibroblasts (PEFs) by coating culture dishes or flasks with 0.1% gelatin in water.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité