Dulbecco's Phosphate Buffered Saline (DPBS), with 2% Fetal Bovine Serum
Fournisseur: STEMCELL Technologies
07905
STMC07905EA 61 EUR InStock
STMC07905
Dulbecco's Phosphate Buffered Saline (DPBS), with 2% Fetal Bovine Serum
Buffers Tampons de culture de tissus
Dulbecco's Phosphate Buffered Saline with 2% fetal bovine serum is recommended for use as a buffer to dilute or wash nucleated cells.
Species: Human, mouse, non human primate, other, rat
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité