Heparin Solution
Fournisseur: STEMCELL Technologies
07980
STMC07980EA 57 EUR InStock
STMC07980
Heparin Solution
Milieux de culture cellulaire
Cell culture supplement.
Heparin is a mucopolysaccharide with anticoagulant properties. It supports the binding of fibroblast growth factor (FGF) to its receptor and increases the stability of FGF. Heparin is used together with epidermal growth factor (EGF) and FGF in NeuroCult™ media for the culture of embryonic rat or adult human neural stem and progenitor cells, and adult mouse neural stem and progenitor cells. For complete instructions, refer to the Technical Manual: In Vitro Proliferation and Differentiation of Neural Stem and Progenitor Cells Using NeuroCult™. Heparin Solution is also required as a supplement in various other STEMCELL culture media, including MammoCult™ Human Medium Kit, EpiCult™-B Mouse Medium Kit, and EC-Cult™-XF Culture Kit.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité