EC-Cult™-XF Culture Kit
Fournisseur: STEMCELL Technologies
08000
STMC08000EA 375 EUR InStock
STMC08000
EC-Cult™-XF Culture Kit
Milieux de culture cellulaire
Culture kit for derivation and proliferation of human endothelial colony-forming cells (ECFCs) and mature endothelial cells.
EC-Cult™-XF Culture Kit is a xeno-free medium for the culture of mature human endothelial cells and endothelial colony-forming cells (ECFCs). It has been developed for the expansion of vascular (large, small, and lymphatic vessel-derived endothelial cells) and perivascular cells (human placental pericytes), as well as for the derivation and proliferation of ECFCs from human umbilical cord blood and peripheral blood.
For complete instructions for the ECFC assay, refer to the Technical Manual: Culture of human endothelial colony-forming cells (ECFCs) using EC‑Cult™-XF culture kit (Document), available at www.stemcell.com or contact us to request a copy. EC-Cult™-XF Medium must be used in conjunction with animal component-free cell attachment substrate (Component; included in EC-Cult™-XF Culture Kit). For passaging, Animal Component-Free Cell Dissociation Kit is required. EC-Cult™-XF Medium must be supplemented with Heparin Solution. Since Heparin Solution contains non-human animal-derived components, the complete medium will not be xeno-free.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité