STEMdiff™ Endothelial Differentiation Kit
Fournisseur: STEMCELL Technologies
08005
STMC08005EA 1054 EUR InStock
STMC08005
STEMdiff™ Endothelial Differentiation Kit
Milieux de culture cellulaire
Serum- and xeno-free kit for differentiation and expansion of human pluripotent stem cell-derived endothelial cells.
STEMdiff™ endothelial differentiation kit includes attachment substrate, animal component-free (ACF) endothelial induction medium, and endothelial expansion medium. It is optimised for the differentiation of human pluripotent stem cells (hPSCs) maintained in mTeSR™1, mTeSR™ plus , or TeSR™-E8™ on Corning® Matrigel® to endothelial-like cells. The kit is designed to be used immediately after early mesoderm induction with STEMdiff™ mesoderm induction medium, available for purchase separately. CollPlant is the manufacturer of the rhCollagen component of the cell attachment substrate. The Product may be used for research purposes only.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité