10% Bovine Serum Albumin in Iscove's MDM
Fournisseur: STEMCELL Technologies
09300
STMC09300EA 1211 EUR InStock
STMC09300
10% Bovine Serum Albumin in Iscove's MDM
Milieux de culture cellulaire
10% BSA in Iscove's MDM.
This product has been developed for use in applications where a culture medium of defined composition is required. It contains pre-screened batches of bovine serum albumin (BSA) that have been selected to support the optimal growth of human hematopoietic progenitor cells in serum-free media formulations. It is also suitable for the culture of mouse hematopoietic progenitor cells in serum-free conditions. Recommended for use with MethoCult™ H4100 or MethoCult™ M3134 for hematopoietic colony-forming unit (CFU) assays (at a final concentration of 1%).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité