HemaTox™ Myeloid Kit
Fournisseur: STEMCELL Technologies
09704
STMC09704EA 1060 EUR InStock
STMC09704
HemaTox™ Myeloid Kit
Milieux de culture cellulaire
Serum-free medium and supplement for the measurement of drug toxicity on myeloid cells.
Screen large numbers of novel drug compounds for their effect on the proliferation and myeloid-specific differentiation of human hematopoietic stem and progenitor cells (HSPCs). Compared to using the colony-forming unit (CFU) assay, the liquid culture-based HemaTox™ assay is more flexible; add test compounds at various points throughout the culture and measure their effects on more primitive or more mature progenitor cells. The HemaTox™ Myeloid Kit promotes the proliferation of CD34+ HSPCs and their differentiation into cells expressing myeloid markers such as CD13, CD14, and CD15. Users can choose the readout method to quantify the response and estimate the IC50 and IC90 of each compound tested.
Test up to 160 conditions - in triplicate - per kit in 5x96-well plates. This kit may be used on its own or in combination with HemaTox™ Erythroid Kit (Catalog #09701) or HemaTox™ Megakaryocyte Kit (Catalog #09707) to assess lineage-specific drug toxicity in parallel.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité