StemSpan™ Leukemic Cell Culture Kit
Fournisseur: STEMCELL Technologies
09720
STMC09720EA 1427 EUR InStock
STMC09720
StemSpan™ Leukemic Cell Culture Kit
Milieux de culture cellulaire
For culture, expansion, and drug screening of chronic and acute myeloid leukemia cells.
StemSpan™ Leukemic Cell Culture Kit has been developed for the in vitro culture and expansion of malignant cells and has been tested on chronic myeloid leukemia (CML) and acute myeloid leukemia (AML) samples. This optimised protocol allows users to expand, culture, and use malignant cells for drug screening. StemSpan™ Leukemic Cell Culture Kit includes the serum-free medium SFEM II, StemSpan™ CD34+ Expansion Supplement (10X), and small molecule UM729.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité