StemSpan™ Lymphoid Differentiation Coating Material (100X)
Fournisseur: STEMCELL Technologies
09925
STMC09925EA 248 EUR InStock
STMC09925
StemSpan™ Lymphoid Differentiation Coating Material (100X)
Milieux de culture cellulaire
Coating material for the differentiation of lymphoid cells.
Use StemSpan™ Lymphoid Differentiation Coating Material (100X) to coat plates for the expansion and differentiation of human CD34+ hematopoietic progenitor cells to T cells. For your convenience, this supplement is available either individually or as part of the StemSpan™ T Cell Generation Kit or the STEMdiff™ T Cell Kit. In this protocol, CD34+ cells are first differentiated to CD7+CD5+ lymphoid progenitor cells, using StemSpan™ Lymphoid Progenitor Expansion Supplement (10X), and then further directed to differentiate to DP T cells. StemSpan™ Lymphoid Differentiation Coating Material (100X) is intended for use in combination with StemSpan™ T Cell Progenitor Maturation Supplement (10X), StemSpan™ SFEM II medium, and StemSpan™ Lymphoid Progenitor Expansion Supplement (10X).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité