StemSpan™ T Cell Generation Kit
Fournisseur: STEMCELL Technologies
09940
STMC09940EA 1600 EUR InStock
STMC09940
StemSpan™ T Cell Generation Kit
Milieux de culture cellulaire
For expansion and differentiation of human CD34+ hematopoietic progenitor cells to T cells.
Use this two-step protocol to reproducibly expand thousands of CD4+CD8+ double-positive (DP) T cells in cultures initiated with CD34+ human cord blood (CB) cells. In step one, CD34+ CB cells are cultured for 14 days in medium containing expansion supplement to promote their proliferation and differentiation into CD7+CD5+ pro-T cells. In step two, pro-T cells generated during those first 14 days are cultured for another four weeks in medium containing maturation supplement to promote their maturation into DP cells. This medium kit is serum- and feeder-free, eliminating variation introduced by serum and stromal cell lines. For convenience, all kit components are also available for purchase individually.
StemSpan™ lymphoid progenitor expansion supplement (10X) contains a combination of recombinant human cytokines and other additives formulated to selectively promote the expansion and differentiation of CD34+ cells isolated from human CB samples to CD7+CD5+ progenitor T (pro-T) cells. Additionally, StemSpan™ T cell progenitor maturation supplement (10X) enables further maturation of pro-T cells to DP cells. It also supports the maturation of DP cells to CD8 single-positive (SP) T cells with additional stimuli. StemSpan™ lymphoid progenitor expansion supplement (10X) and StemSpan™ T cell progenitor maturation supplement (10X) are intended for use in combination with StemSpan™ SFEM II medium, and on plates coated with StemSpan™ lymphoid differentiation coating material (100X). An optional protocol is available to further mature DP cells to CD8 SP T cells. This extended protocol uses a complete T cell progenitor maturation medium made with reagents included in the StemSpan™ T cell generation kit and must be combined with ImmunoCult™ CD3/CD28/CD2 T cell activator and IL-15.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité