StemSpan™ NK Cell Differentiation Supplement (100X)
Fournisseur: STEMCELL Technologies
09950
STMC09950EA 250 EUR InStock
STMC09950
StemSpan™ NK Cell Differentiation Supplement (100X)
Milieux de culture cellulaire
Supplement for the differentiation of lymphoid progenitors to NK cells.
Support the differentiation of lymphoid progenitor cells to natural killer (NK) cells with StemSpan™ NK Cell Differentiation Supplement (100X). For convenience, the supplement is available either individually or as part of the StemSpan™ NK Cell Generation Kit or the STEMdiff™ NK Cell Kit. Lymphoid progenitor cells generated by differentiation of CD34+ cells in StemSpan™ SFEM II medium containing StemSpan™ Lymphoid Progenitor Expansion Supplement (10X) on plates coated with StemSpan™ Lymphoid Differentiation Coating Material (100X) can be further differentiated to NK cells using this supplement. The StemSpan™ NK Cell Differentiation Supplement (100X) is intended to be used in combination with StemSpan™ SFEM II and UM729 to promote differentiation into CD56+ NK cells.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité