STEMdiff™ Astrocyte Maturation Kit
Fournisseur: STEMCELL Technologies
100-0016
STMC100-0016EA 336 EUR InStock
STMC100-0016
STEMdiff™ Astrocyte Maturation Kit
Milieux de culture cellulaire
Maturation kit for the generation of astrocytes from human ES and iPS cell-derived astrocyte precursors.
STEMdiff™ Astrocyte Maturation Kit is used to rapidly and efficiently generate cortical-type astrocytes from astrocytic precursors derived from human pluripotent stem cells (hPSCs) using the STEMdiff™ Astrocyte Differentiation Kit. Using this system, a highly pure population of astrocytes (an average of >70% S100B-positive and >60% GFAP-positive astrocytes; <15% doublecortin-positive neurons) can be generated from hPSCs in as few as 7 weeks and can be maintained long term in culture. Cells derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité