STEMdiff™ Microglia Maturation Kit
Fournisseur: STEMCELL Technologies
100-0020
STMC100-0020EA 928 EUR InStock
STMC100-0020
STEMdiff™ Microglia Maturation Kit
Milieux de culture cellulaire
Maturation kit for the generation of microglia from human ES and iPS cell-derived microglia precursors.
The STEMdiff™ microglia culture system comprises STEMdiff™ microglia differentiation kit and STEMdiff™ microglia maturation kit. Together, these kits are used to differentiate and mature microglia derived from human pluripotent stem cells (hPSCs) using STEMdiff™ hematopoietic kit. Based on the protocol from the laboratory, the resulting cells are a highly pure population of microglia (>80% CD45/CD11b-positive, >50% TREM2-positive microglia; >20% morphologically distinct monocytes or macrophages). Cells derived using these products are versatile tools for modeling neuroinflammation, studying human neurological development and disease, co-culture applications, and toxicity testing.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité