STEMdiff™ Midbrain Neuron Differentiation Kit
Fournisseur: STEMCELL Technologies
100-0038
STMC100-0038EA 391 EUR InStock
STMC100-0038
STEMdiff™ Midbrain Neuron Differentiation Kit
Milieux de culture cellulaire
Differentiation kit for the generation of midbrain neuronal precursors from human ES and iPS cell-derived neural progenitor cells.
STEMdiff™ Midbrain Neuron Differentiation Kit is used to generate midbrain neuronal precursors from neural progenitor cells (NPCs) derived from human pluripotent stem cells (hPSCs) using STEMdiff™ SMADi Neural Induction Kit. The midbrain neuronal precursors are further matured into midbrain neurons using STEMdiff™ Midbrain Maturation Kit. These media will produce a population of midbrain neurons (≥15% TH-positive dopaminergic neurons; ≥90% class III βtubulin-positive neurons; <10% GFAP-positive astrocytes). Cells derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité