EasySep™ Release Human PSC-Derived Neural Crest Cell Positive Selection Kit
Fournisseur: STEMCELL Technologies
100-0047
STMC100-0047EA 960 EUR InStock
STMC100-0047
EasySep™ Release Human PSC-Derived Neural Crest Cell Positive Selection Kit
Cell Separation Media
Immunomagnetic positive selection kit using particle release technology
  • Highly purified human PSC-derived neural crest cells isolated in less than 20 minutes
  • No-wash removal of EasySep™ Releasable RapidSpheres™
  • Compatible with downstream differentiation protocols
The EasySep™ Release Human PSC-Derived Neural Crest Cell Positive Selection Kit is designed to isolate CD271⁺ cells from human pluripotent stem cell (PSC)-derived neural crest cell culture by immunomagnetic positive selection.
Desired cells are labelled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated CD271+ cells, which are immediately available for downstream applications.
Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands.
Attention: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité