EasySep™ Release Human PSC-Derived Neural Crest Cell Positive Selection Kit
Fournisseur: STEMCELL Technologies
100-0047
STMC100-0047EA 960 EUR InStock
STMC100-0047
EasySep™ Release Human PSC-Derived Neural Crest Cell Positive Selection Kit
Milieux de séparation cellulaire
Immunomagnetic positive selection kit using particle release technology
  • Highly purified human PSC-derived neural crest cells isolated in less than 20 minutes
  • No-wash removal of EasySep™ Releasable RapidSpheres™
  • Compatible with downstream differentiation protocols
The EasySep™ Release Human PSC-Derived Neural Crest Cell Positive Selection Kit is designed to isolate CD271⁺ cells from human pluripotent stem cell (PSC)-derived neural crest cell culture by immunomagnetic positive selection.
Desired cells are labelled with antibodies and magneticMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité