StemSpan™-XF Medium
Fournisseur: STEMCELL Technologies
100-0073
STMC100-0073EA 695 EUR InStock
STMC100-0073
StemSpan™-XF Medium
Milieux de culture cellulaire
Xeno-free and serum-free medium for culture and expansion of human hematopoietic cells.
StemSpan™-XF has been developed for the in vitro culture and expansion of human hematopoietic cells, when the appropriate growth factors and supplements are added. This allows users the flexibility to prepare medium that meets their requirements. StemSpan™-XF contains pre-tested human-derived and recombinant human (rh) proteins. Using appropriate StemSpan™ expansion supplements, StemSpan™-XF may be used to expand CD34+ cells isolated from human cord blood, mobilised peripheral blood, or bone marrow samples, or to expand and differentiate lineage-committed progenitor cells to generate populations of erythroid, myeloid, or megakaryocyte progenitor cells.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité