Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD63 Mouse Monoclonal Antibody [clone: H5C6]

Fournisseur: STEMCELL Technologies
100-0139
STMC100-0139EA 521 EUR InStock
STMC100-0139
Anti-CD63 Mouse Monoclonal Antibody [clone: H5C6]
Anticorps
Mouse monoclonal antibody against human, rhesus, cynomolgus CD63, unconjugated.

The H5C6 antibody reacts with an extracellular epitope on human CD63, a 30-60 kDa type III lysosomal glycoprotein and member of the tetraspanin family. CD63 exhibits broad expression, including on the surface of monocytes, macrophages, activated basophils, fibroblasts, smooth muscle cells, and activated platelets. CD63 is found in late endosomes, lysosomes, and secretory vesicles, and cycles among these compartments. It is also a marker characteristic of extracellular vesicles. Tetraspanins like CD63 contain four transmembrane domains, two extracellular loops, and short cytoplasmic N- and C-termini. CD63 associates with several integrins, co-receptors, and other proteins to form multimolecular complexes in the plasma membrane called tetraspanin-enriched microdomains. The protein is involved in several cellular processes, including cell activation, adhesion, differentiation, and tumor invasion. CD63 has been implicated in tumor progression, and a deficiency of the protein is associated with Hermansky-Pudlak syndrome, a rare autosomal recessive disorder presenting with platelet dysfunction and defects in lysosomal storage.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD63
Clonality: Monoclonal
Clone: H5C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: African green monkey;Baboon;Capuchin monkey;Chimpanzee;Cynomolgus monkey;Human;Rhesus monkey

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité