STEMdiff™ Myogenic Progenitor Supplement Kit
Fournisseur: STEMCELL Technologies
100-0151
STMC100-0151EA 756 EUR InStock
STMC100-0151
STEMdiff™ Myogenic Progenitor Supplement Kit
Milieux de culture cellulaire
Serum-free supplements for differentiation of human pluripotent stem cells (hPSCs) to myogenic progenitor cells.
STEMdiff™ Myogenic Progenitor Supplement Kit comprises serum-free supplements for the differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells to myogenic progenitor cells. These cells, which are characterised by myogenic cell markers such as CD56 and CD82, can be culture-expanded for 5+ passages using MyoCult™-SF Expansion Supplement Kit and further differentiated to functional multinucleated MyHC+ myotubes at high efficiency using MyoCult™ Differentiation Kit. These myotubes can be used in various downstream applications and analyses. This kit is compatible with human ES and iPS cells maintained in mTeSR™ 1, mTeSR™ Plus, or TeSR™-E8™.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité