STEMdiff™ NK Cell Kit
Fournisseur: STEMCELL Technologies
100-0170
STMC100-0170EA 1468 EUR InStock
STMC100-0170
STEMdiff™ NK Cell Kit
Milieux de culture cellulaire
For expansion and differentiation of hPSCs to NK cells.
Differentiate human pluripotent stem cells (hPSCs) to NK cells with the feeder-free and serum-free STEMdiff™ NK Cell Kit. The STEMdiff™ NK Cell Kit protocol first uses animal component-free STEMdiff™ Hematopoietic - EB reagents for the generation of embryoid bodies (EBs) from hPSCs which are then further differentiated into CD34+ cells.
The kit includes all the EB reagents necessary for this step:
STEMdiff™ Hematopoietic - EB Basal Medium
STEMdiff™ Hematopoietic - EB Supplement A
STEMdiff™ Hematopoietic - EB Supplement B
The CD34+ cells are then further directed to differentiate to CD56+ NK cells using StemSpan™ reagents:
StemSpan™ SFEM II
StemSpan™ Lymphoid Progenitor Expansion Supplement (10X)
StemSpan™ Lymphoid Differentiation Coating Material (100X)
StemSpan™ NK Cell Differentiation Supplement (100X)
For convenience, the STEMdiff™ Hematopoietic - EB and StemSpan™ reagents included with the kit are also available individually.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité