STEMdiff™ Hematopoietic - EB Supplement B
Fournisseur: STEMCELL Technologies
100-0173
STMC100-0173EA 390 EUR InStock
STMC100-0173
STEMdiff™ Hematopoietic - EB Supplement B
Milieux de culture cellulaire
Supplement for hematopoietic differentiation optimised for lymphoid potential.
Use STEMdiff™ Hematopoietic - EB Supplement B in combination with STEMdiff™ Hematopoietic - EB Basal Medium and STEMdiff™ Hematopoietic - EB Supplement A to facilitate hematopoietic differentiation during the generation of human pluripotent stem cell (hPSC)-derived CD34+ cells. Available either individually or as part of the STEMdiff™ NK Cell Kit or STEMdiff™ T Cell Kit, where it can be used to promote the differentiation of hPSCs to NK or T cells, respectively. For detailed information on the two differentiation protocols, please explore the STEMdiff™ NK Cell Kit Technical Manual or the STEMdiff™ T Cell Kit Technical Manual.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité