Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-CD81 (TAPA-1) Mouse Monoclonal Antibody [clone: 5A6]

Fournisseur: STEMCELL Technologies
100-0209
STMC100-0209EA 321 EUR InStock
STMC100-0209
Anti-CD81 (TAPA-1) Mouse Monoclonal Antibody [clone: 5A6]
Anticorps
Mouse monoclonal antibody against human, rhesus, cynomolgus CD81, unconjugated.

The 5A6 antibody reacts with an epitope located in the large extracellular loop (EC2) of human CD81 (TAPA-1). CD81 exhibits broad expression, including on the surface of B and T cells, NK cells, monocytes, dendritic cells, thymocytes, endothelial cells, and fibroblasts. It is also a marker characteristic of extracellular vesicles. CD81, a 26 kDa non-glycosylated type III transmembrane protein, belongs to the tetraspanin family. Tetraspanins contain four transmembrane domains, two extracellular loops, and short cytoplasmic N- and C-termini. CD81 associates with numerous integrins, co-receptors, and other proteins to form multimolecular complexes in the plasma membrane called tetraspanin-enriched microdomains. Palmitoylation of the juxtamembrane cysteine residues is important for its association with other proteins and the plasma membrane. CD81 modulates a diverse array of biological functions including cell-cell adhesion and fusion, cell migration, and proliferation. For example, together with CD19 and CD21, CD81 forms a co-receptor for the B cell receptor on B cells. The epitope of the 5A6 antibody is hidden when CD81 is in complex with CD19 but accessible following activation of the B cell. Mutations in CD81 have been associated with the immune disorder common variable immunodeficiency (CVID). CD81 has also been implicated in mediating the entry of hepatitis C virus into cells.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: CD81 (TAPA-1)
Clonality: Monoclonal
Clone: 5A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: African green monkey, Baboon, Chimpanzee, Cynomolgus monkey, Human, Rhesus monkey

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité