STEMdiff™ Lung Progenitor Kit
Fournisseur: STEMCELL Technologies
100-0230
STMC100-0230EA 1091 EUR InStock
STMC100-0230
STEMdiff™ Lung Progenitor Kit
Milieux de culture cellulaire
Serum-free medium for efficient and reproducible generation of lung progenitor cells from human ES and iPS cells.
STEMdiff™ Lung Progenitor Kit is a serum-free culture medium system for efficient and reproducible generation of lung progenitor cells from human embryonic stem (ES) and induced pluripotent stem (iPS) cells through three stages of differentiation: 1) definitive endoderm, 2) anterior foregut endoderm, and 3) lung progenitor cells. Differentiated cells will express NKX2.1, a key marker of lung progenitor cells. The resulting cells can be further matured towards proximal or distal airway cells using published protocols for the study of lung diseases and lung development.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité