Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody [clone: M5/114.15.2]

Fournisseur: STEMCELL Technologies
100-0297
STMC100-0297EA 123 EUR InStock
STMC100-0297
Anti-MHC Class II (I-A/I-E) Rat Monoclonal Antibody [clone: M5/114.15.2]
Anticorps
Rat monoclonal IgG2b, antibody against mouse MHC Class II (I-A/I-E), unconjugated.

  • B cells
  • Macrophages
  • Monocytes
  • T cells

The M5/114.15.2 (M5/114) antibody reacts with an extracellular epitope on the mouse major histocompatibility complex class II receptor (MHC-II). MHC-II is a heterodimeric transmembrane glycoprotein comprising an α subunit associated non-covalently with a β subunit. In mice, MHC-II complexes are designated I-A and I-E and comprise Aα/Aβ and Eα/Eβ heterodimers, respectively. Together with the CD3/T cell receptor (TCR) complex and CD4 molecules, MHC-II mediates a critical function in presenting peptides generated from hydrolysis of exogenous antigens by antigen-presenting cells to CD4+ T (helper) cells, thereby either suppressing or inducing an immune response to the peptides. The M5/114.15.2 antibody recognizes a polymorphic determinant shared by the I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek (but not I-Af, I-Ak, or I-As) MHC class II alloantigens expressed on B cells, dendritic cells, macrophages, monocytes, and activated T cells of mice carrying the H-2b, d, p, q, r, and u haplotypes (but not haplotypes H-2f or H-2s). The M5/114 antibody reportedly inhibits I-A-restricted responses by T cells of the H-2b, d, q, and u (but not H-2f, k, or s) haplotypes. It does not react with cells from NOD (H-2g7) mice.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: MHC Class II (I-A/I-E)
Clonality: Monoclonal
Clone: M5/114.15.2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité