Annexin V Binding Buffer
Fournisseur: STEMCELL Technologies
100-0334
STMC100-0334EA 75 EUR InStock
STMC100-0334
Annexin V Binding Buffer
Systèmes de liaison et de marquage des protéines
Assay buffer for labeling with Annexin V - 50 ml.
Annexin V Binding Buffer is an isotonic buffer used in conjunction with Annexin V dyes (e.g. Catalog #100-0329) and either 7-AAD (Catalog #75001) or Propidium Iodide (Catalog #75002). This buffer contains an optimal concentration of calcium that facilitates the binding of Annexin V to phosphatidylserine on the cell surface, which is essential for the detection of early-stage cell apoptosis by flow cytometry.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité