STEMdiff™ Sensory Neuron Differentiation Kit
Fournisseur: STEMCELL Technologies
100-0341
STMC100-0341EA 282 EUR InStock
STMC100-0341
STEMdiff™ Sensory Neuron Differentiation Kit
Milieux de culture cellulaire
Differentiation kit for the generation of sensory neuronal precursors from human iPS cell-derived neural crest cells.
Generate functional, human-specific sensory neurons in a dish with this easy-to-use culture system. After obtaining pluripotent stem cell (PSC)-derived neural crest cells (NCCs) with STEMdiff™ Neural Crest Differentiation Kit, use the serum-free STEMdiff™ Sensory Neuron Differentiation Kit to generate sensory neuron precursors. STEMdiff™ Sensory Neuron Maturation Kit can then be used to maintain and mature the sensory neuron precursors.
The resulting cells are BRN3A-positive and more than 70% TUJ1-positive. With the physiological glucose and osmolarity conditions provided by BrainPhys™ as the basal medium, the neurons exhibit activity in response to sensory ligands and changes in temperature. The generated population of functional human sensory neurons can be used for drug discovery and pain research applications.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité