ClonaCell™ CHO AOF Supplement
Fournisseur: STEMCELL Technologies
100-0382
STMC100-0382EA 176 EUR InStock
STMC100-0382
ClonaCell™ CHO AOF Supplement
Milieux de culture cellulaire
Animal origin-free, serum-free, L-glutamine-free, and phenol red-free 40X culture supplement for cloning during CHO cell line development.
Save time and precious clones during cell line development by increasing cloning efficiency and promoting robust growth of Chinese hamster ovary (CHO) cells with ClonaCell™-CHO AOF Supplement in the base medium. Formulated without phenol red, this supplement offers precipitate-free culture for improved imaging clarity and visualisation of clonality, compared to ClonaCell™-CHO ACF Supplement. This animal origin-free (AOF) supplement is formulated with no animal-derived raw materials to the secondary level of manufacturing. It contains recombinant proteins and chemically defined components and is free of serum, hydrolysates, L-glutamine, or selection agents. Use this 40X supplement with a variety of base media suitable for CHO cell culture, including protein-free ClonaCell™-CHO CD Liquid Medium or semi-solid ClonaCell™-CHO CD Medium. Explore liquid and semi-solid media for efficient cell cloning and cell line generation in the ClonaCell™ brand page.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité