HepatiCult™ Organoid Initiation Medium (Human)
Fournisseur: STEMCELL Technologies
100-0384
STMC100-0384EA 529 EUR InStock
STMC100-0384
HepatiCult™ Organoid Initiation Medium (Human)
Milieux de culture cellulaire
Culture medium to initiate organoids from primary liver tissue.
Efficiently generate physiologically relevant human liver organoids across different donor lines with HepatiCult™ Organoid Initiation Medium (OIM) (Human). Available either individually or as part of HepatiCult™ organoid kit (Human), HepatiCult™ OIM can be used to generate hepatic organoids from fresh or cryopreserved human liver tissue. These organoids may then be further expanded and maintained in HepatiCult™ organoid growth medium (Human) and subsequently differentiated with HepatiCult™ organoid differentiation medium (Human) to generate mature organoids that demonstrate hepatic functionality, including CYP3A4 activity.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité