STEMdiff™ Gastric Organoid Differentiation Kit
Fournisseur: STEMCELL Technologies
100-0475
STMC100-0475EA 1213 EUR InStock
STMC100-0475
STEMdiff™ Gastric Organoid Differentiation Kit
Milieux de culture cellulaire
Culture medium kit for differentiation of human gastric organoids.
Reliably generate hPSC-derived gastric organoids for studying gastric development, inflammation, regeneration, microbial interaction, or for disease modeling. With the STEMdiff™ Gastric Organoid Differentiation Kit, you can grow and expand organoids across hPSC lines with high efficiency and reproducibility, to form a convenient model system with direct relevance to the developing stomach. This kit’s simple, four-stage protocol, based on publications by McCracken et al. (Nature 2014) and Bartfeld et al. (Gastroenterology 2015), enables the generation and differentiation of gastric organoids directly from hPSCs as well as the long-term expansion and cryopreservation of gastric organoids. Organoids cultured with this kit exhibit a cellular composition and organisation that recapitulates that of the developing gastric epithelium and associated mesenchyme, providing a physiologically relevant model system for gastric research, as well as drug discovery and compound screening. This kit is optimised for differentiation of cells maintained in mTeSR™1 or mTeSR™ Plus.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité