STEMdiff™ Gastric Organoid Expansion Medium
Fournisseur: STEMCELL Technologies
100-0490
STMC100-0490EA 556 EUR InStock
STMC100-0490
STEMdiff™ Gastric Organoid Expansion Medium
Milieux de culture cellulaire
Culture medium kit for expansion of human gastric organoids.
Reliably and efficiently expand human pluripotent stem cell (hPSC)-derived gastric organoids to use as a model system with direct relevance to the developing stomach. With serum-free STEMdiff™ Gastric Organoid Expansion Medium, you can reproducibly maintain gastric organoid cultures for studying gastric development, inflammation, regeneration, and microbial interaction, or for disease modeling. Based on publications by McCracken et al. and Bartfeld et al., STEMdiff™ Gastric Organoid Expansion Medium is available as part of STEMdiff™ Gastric Organoid Differentiation Kit for the generation and differentiation of gastric hPSC-derived organoids, as well as individually for long-term maintenance of the resulting cultures.
Gastric organoids generated using the differentiation kit exhibit a cellular composition and organization that recapitulates that of the developing gastric epithelium and associated mesenchyme, providing a physiologically relevant model system for gastric research, as well as drug discovery and compound screening. Use STEMdiff™ Gastric Organoid Expansion Medium at the end of the four-stage differentiation protocol to efficiently expand these hPSC-derived gastric organoids for use in downstream applications or for cryopreservation.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité