Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Anti-SARS-CoV-2 Spike Protein S1 Receptor Binding Domain Human Monoclonal Antibody [clone: Covi-2]

Fournisseur: STEMCELL Technologies
100-0584
STMC100-0584EA 496 EUR InStock
STMC100-0584
Anti-SARS-CoV-2 Spike Protein S1 Receptor Binding Domain Human Monoclonal Antibody [clone: Covi-2]
Anticorps
Human monoclonal IgG1 antibody against SARS-CoV-2 (2019-nCoV) S protein (HEK293-expressed recombinant).

The Anti-SARS-CoV-2 Spike Protein S1 Receptor-Binding Domain Antibody, Clone Covi-2 (Blocking/Recombinant antibody reacts with Spike Protein S1 receptor binding domain (RBD) expressed by SARS-associated coronavirus 2 (SARS-CoV-2/2019-nCoV). The Spike (S) protein is a type I transmembrane glycoprotein present on the surface of coronaviruses (CoV). Entry of CoV into host cells is mediated by the S protein, where it interacts with the cell-surface receptor angiotensin-converting enzyme 2 (ACE2). In humans, ACE2 is expressed in several organs and tissues, including intestinal and respiratory epithelium. The S protein has two subunits, S1 and S2, where S1 primarily consists of the 193 amino acid long RBD and the N-terminal domain (NTD). The S2 domain is responsible for membrane fusion. During CoV infection, the S protein is cleaved into the N-terminal S1 subunit and C-terminal S2 subunit by host proteases, transforming its conformation from the pre-fusion to the post-fusion state. The S protein has been shown to play a key role in the induction of neutralizing antibody and T cell responses, which may lead to protective immunity. The RBD binds to ACE2, while the function of the NTD is not well understood. The Covi-2 antibody blocks the interaction of the S1 subunit with the ACE2 receptor and binds to a different epitope than the Covi-1 antibody.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: SARS-CoV-2 Spike Protein S1 Receptor Binding Domain
Clonality: Monoclonal
Clone: Covi-2
Conjugation:
Epitope:
Host: Human
Isotype: IgG1
Reactivity:

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité