STEMdiff™ Blood Vessel Organoid Kit
Fournisseur: STEMCELL Technologies
100-0651
STMC100-0651EA 1310 EUR InStock
STMC100-0651
STEMdiff™ Blood Vessel Organoid Kit
Milieux de culture cellulaire
Media and supplements for differentiation of human pluripotent stem cells into blood vessels organoids.
Generate physiologically relevant blood vessel organoids - easily and with high yield - for tissue repair and regeneration studies or to model vascular disease in vitro by using STEMdiff™ Blood Vessel Organoid Kit. Use STEMdiff™ Blood Vessel Organoid Kit for efficient and reproducible generation of blood vessel organoids from human pluripotent stem cells (hPSCs), with the option to scale up for high-throughput screening in a 96-well format. Compared to homemade organoid culture media formulations, organoids generated with this easy-to-use kit develop extensive and physiologically relevant vascular networks and strong endothelial cell-pericyte interactions.
Blood vessel organoids generated using this kit have CD31+/CD34+/CD144+/KDR+ endothelial cells and PDGFR-β+/CD146+/SMA+/NG-2+ pericytes.The simple protocol also eliminates the need for serum screening and labor-intensive protocol optimisation. Use STEMdiff™ Blood Vessel Organoid Kit with mTeSR™ Plus or mTeSR™1 culture media for an easy transition from hPSCs to organoids. For convenience, STEMdiff™ Blood Vessel Organoid Maturation Medium, required for the maturation of hPSC-derived blood vessel organoids, is available either as a part of the kit or for individual sale. To differentiate hPSCs to endothelial cells, use our STEMdiff™ Endothelial Differentiation Kit.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité