STEMdiff™ Blood Vessel Organoid Maturation Medium
Fournisseur: STEMCELL Technologies
100-0658
STMC100-0658EA 734 EUR InStock
STMC100-0658
STEMdiff™ Blood Vessel Organoid Maturation Medium
Milieux de culture cellulaire
Media for maturation of human pluripotent stem cells derived blood vessels organoids.
Use STEMdiff™ Blood Vessel Organoid Maturation Medium to expand and grow mature blood vessel organoids generated using the STEMdiff™ Blood Vessel Organoid Kit or homemade protocols, with the option to scale up for high-throughput screening in a 96-well format. Compared to homemade organoid culture media formulations, organoids generated with this easy-to-use kit develop extensive and physiologically relevant vascular networks and strong endothelial cell-pericyte interactions. Blood vessel organoids matured using this medium have CD31+/CD34+/CD144+/KDR+ endothelial cells and PDGFR-β+/CD146+/SMA+/NG-2+ pericytes. The simple protocol also eliminates the need for serum screening and labor-intensive protocol optimization.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité