SARS-CoV-2 (NS8) Peptide Pool
Fournisseur: STEMCELL Technologies
100-0663
STMC100-0663EA 354 EUR InStock
STMC100-0663
SARS-CoV-2 (NS8) Peptide Pool
Protéines et Peptides
The SARS-CoV-2 (NS8) Peptide pool is a lyophilised mixture of 28 peptides from non-structural protein 8 (NS8) of SARS-CoV-2. NS8 has been identified as one of the proteins exclusively conserved in a few species of the coronavirus family, and it contains Class II MHC epitopes. The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 121 on NS8.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité