EBV (EBNA-1) Peptide Pool
Fournisseur: STEMCELL Technologies
100-0669
STMC100-0669EA 447 EUR InStock
STMC100-0669
EBV (EBNA-1) Peptide Pool
Protéines et Peptides
The EBV (EBNA-1) Peptide pool is a lyophilised mixture of 158 peptides from Epstein-Barr nuclear antigen 1 (EBNA-1) of Epstein-Barr virus (EBV; strain B95-8). EBNA-1 plays a critical role in the stable latent infection by EBV and may contribute to the survival of EBV-infected cells through its interaction with ubiquitin carboxyl-terminal hydrolase 7 (USP7). The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 641 on EBNA-1.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité