EBV (LMP2) Peptide Pool
Fournisseur: STEMCELL Technologies
100-0671
STMC100-0671EA 531 EUR InStock
STMC100-0671
EBV (LMP2) Peptide Pool
Protéines et Peptides
The EBV (LMP2) Peptide pool is a lyophilised mixture of 122 peptides from latent membrane protein 2 (LMP2) of Epstein-Barr virus (EBV; strain B95-8). LMP2 is expressed in EBV latency, as well as in EBV-associated pathologies and it blocks reactivation of EBV from latency. The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 497 on LMP2.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité