CEF (HLA Class I Control) Peptide Pool
Fournisseur: STEMCELL Technologies
100-0675
STMC100-0675EA 194 EUR InStock
STMC100-0675
CEF (HLA Class I Control) Peptide Pool
Protéines et Peptides
The CEF (HLA Class I Control) Peptide pool is a lyophilised mixture of 32 peptides from cytomegalovirus (CMV), Epstein-Barr virus (EBV), and influenza virus. The pool consists of defined HLA class I-restricted T cell epitopes from these three viruses, and can be used as a positive control to stimulate T cells.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité