SARS-CoV-2 (Spike Protein) Peptide Pool
Fournisseur: STEMCELL Technologies
100-0676
STMC100-0676EA 868 EUR InStock
STMC100-0676
SARS-CoV-2 (Spike Protein) Peptide Pool
Protéines et Peptides
The SARS-CoV-2 (spike protein) peptide pool is provided as two lyophilised mixtures (subpools) from the SARS-CoV-2 spike protein. Each subpool contains 158 peptides, for a total of 316 peptides. The virus attaches to the cell membrane of the host through the interaction between spike protein and angiotensin-converting enzyme 2 (ACE2) receptor, and the spike protein plays a critical role in viral entry. The subpools consist of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 1273 on the spike protein.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité