STEMdiff™ Sensory Neuron Maturation Kit
Fournisseur: STEMCELL Technologies
100-0684
STMC100-0684EA 330 EUR InStock
STMC100-0684
STEMdiff™ Sensory Neuron Maturation Kit
Milieux de culture cellulaire
Maturation kit for generation of functional sensory neurons from human iPS cell-derived neuronal precursor cells.
Generate functional, human-specific sensory neurons in a dish with this easy-to-use culture system. STEMdiff™ Sensory Neuron Maturation Kit is the final step in a workflow beginning from human pluripotent stem cells (hPSCs) and leading to a population of sensory neurons that can be used for drug discovery and pain research applications. After obtaining PSC-derived neural crest cells (NCCs) with STEMdiff™ neural crest differentiation kit, use the serum-free STEMdiff™ sensory neuron differentiation Kit to generate sensory neuron precursors. STEMdiff™ sensory neuron maturation kit can then be used to maintain and mature the sensory neuron precursors. The resulting cells are BRN3A-positive and more than 70% TUJ1-positive. With the physiological glucose and osmolarity conditions provided by BrainPhys™ as the basal medium, the neurons exhibit activity in response to sensory ligands and changes in temperature.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité