Human SARS-CoV-2 Nucleoprotein IgG Antibody ELISA Kit
Fournisseur: STEMCELL Technologies
100-0686
STMC100-0686EA 1048 EUR InStock
STMC100-0686
Human SARS-CoV-2 Nucleoprotein IgG Antibody ELISA Kit
Tests Tests ELISA
For the detection of human anti-SARS-CoV-2 nucleoprotein IgG antibodies.
The Human SARS-CoV-2 Nucleoprotein IgG Antibody ELISA Kit is designed for the qualitative detection of human SARS-CoV-2 nucleoprotein (NP) IgG antibody in biological fluids such as serum and plasma. Coronaviruses (CoV), including SARS-CoV-2, are enveloped and contain single-stranded (positive-sense) RNA associated with a nucleoprotein.
These nucleoproteins are abundantly produced within CoV-infected cells, where their primary function is to package the viral genome into flexible, helical ribonucleoprotein complexes called nucleocapsids. The nucleocapsid binds to genomic RNA and is understood to interact with viral membrane proteins during virion assembly, therefore playing a critical role in ensuring the efficiency of viral transcription, translation, and assembly.
In host cells, the nucleocapsid can directly impact normal cell functions and cause cell-cycle deregulation, increased COX-2 production, and inhibition of interferon production. The assay is based on the indirect ELISA method, in which samples are added to ELISA strip plates pre-coated with human novel coronavirus nucleoprotein.
Antibodies present in the sample specific for the coronavirus nucleoprotein will bind to the pre-coated antigen. The captured antibodies are detected by addition of an anti-human IgG-conjugated horseradish peroxidase (HRP).
Addition of the chromogenic enzyme substrate 3,3’,5,5’ tetramethylbenzidine (TMB) results in a coloured product with an intensity directly proportional to the concentration of human novel coronavirus nucleoprotein IgG antibody in the sample.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité