ImmunoCult™ NK Cell Expansion Kit
Fournisseur: STEMCELL Technologies
100-0711
STMC100-0711EA 662 EUR InStock
STMC100-0711
ImmunoCult™ NK Cell Expansion Kit
Milieux de culture cellulaire
Culture medium kit for expansion of human NK cells.
Consistently expand natural killer (NK) cells in serum-free conditions without the use of problematic feeder cells. Use ImmunoCult™ NK Cell Expansion Kit to provide optimised culture conditions for expanding NK cells with high yields. This kit comprises ImmunoCult™ NK Cell Base Medium, ImmunoCult™ NK Cell Expansion Supplement, and ImmunoCult™ NK Cell Expansion Coating Material, providing a complete, easy-to-use culture system. After just 14 days in culture, cells can be harvested and used directly in downstream applications. This kit is compatible with many of our other upstream and downstream products. For example, use EasySep™ cell separation kits to isolate NK cells that can then be immediately expanded using ImmunoCult™ NK Cell Expansion Kit.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité