LysoBrite™ Lysosome Dye, Deep Red
Fournisseur: STEMCELL Technologies
100-0819
STMC100-0819EA 324 EUR InStock
STMC100-0819
LysoBrite™ Lysosome Dye, Deep Red
Systèmes de liaison et de marquage des protéines
Fluorescent dyes used to label and track lysosomes in live cells - 500 tests.
LysoBrite™ dyes are fluorescent dyes that selectively stain lysosomes and other acidic organelles. Due to their hydrophobic nature, LysoBrite™ dyes permeate live cells and selectively accumulate in the lysosomes via the lysosome’s pH gradient. The fluorescence property of these dyes significantly increases within the lysosome’s acidic environment; this fluorescence can be measured using flow cytometry and fluorescence imaging. Due to their selective fluorescence inside lysosomes, LysoBrite™ dyes provide low background and high contrast during imaging. They can be used with proliferating and non-proliferating cells, and are suitable for suspension and adherent cells.
CAS. number: 67-68-5
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité