L'expérience en ligne d'Avantor évolue pour mieux vous accompagner ! À compter du 4 avril 2025, nos clients seront migrés vers une nouvelle plateforme pour une expérience d'achat en ligne simplifiée.
Phalloidin, iFluor™ 647
Fournisseur: STEMCELL Technologies
100-0854
STMC100-0854EA 335 EUR InStock
STMC100-0854
Phalloidin, iFluor™ 647
Protéines et Peptides
Fluorescent dye conjugates for detection of F-actin in cell culture experiments - 300 tests.
Phalloidin is a heptapeptide derived from the poisonous mushroom Amanita phalloides. Phalloidin binds with a high specificity to F-actin filaments in cells, preventing filament depolymerisation. Fluorescently labeled phalloidin can be used in labelling, identification, and quantitation of F-actin in formaldehyde-fixed and permeabilised cells. Fluorescently labeled phalloidin has lower non-specific binding compared to actin antibodies across most species of plants and animals, which provides low background and high contrast during imaging.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité