Human Recombinant TGF-beta 1 (Cell Culture Ready)
Fournisseur: STEMCELL Technologies
100-0856
STMC100-0856EA 879 EUR InStock
STMC100-0856
Human Recombinant TGF-beta 1 (Cell Culture Ready)
Protéines et Peptides
Conveniently and consistently add transforming growth factor beta 1 (TGF-ꞵ1) to your cultures with this cell culture-ready formulation. This human recombinant TGF-ꞵ1 is precisely reconstituted to 0,1 mg/ml in 100 mM acetic acid and requires no additional preparation, improving reproducibility. Endotoxin levels are verified, using the LAL method to ensure consistency for use across multiple applications. A member of the TGF-β superfamily, TGF-β1 regulates diverse cellular phenotypes. TGF-β1 binds to serine-threonine kinase type I and II receptors and activates signal transduction through SMAD2/3 proteins.
TGF-β1 has been used for:
hPSC culture maintenance:
Use TGF-β1 together with basic fibroblast growth factor (bFGF) to support the culture of undifferentiated human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells (Amit et al. Biol Reprod, 2004).
Exogenus cell signaling:
Regulate a variety of functions, including cell proliferation, differentiation, and metabolism (Massagué Nat Rev Mol Cell Biol, 2000; McDowell et al. Curr Biol, 1997).
Immune cell Homeostatis:
Modulate B cell responses and mediate immunosuppressive effects on T cells and neutrophils (Letterio and Roberts Annu Rev Immunol, 1998).
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité